Order Entry
Ireland
ContactUsLinkComponent
Ethyl-4-hydroxy-3-methoxycinnamate 98%
Ethyl-4-hydroxy-3-methoxycinnamate 98%
Catalog # APOSOR24814-25G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-4-hydroxy-3-methoxycinnamate 98%
Catalog # APOSOR24814-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-hydroxy-3-methoxyphenyl)acrylate, Ethyl Ferulate, Ferulic Acid Ethyl Ester
 :
 :222.24 g/mol
Boiling Pt:164…166 °C/0,5mmHg
Melting Pt:63…65 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:4046-02-0
Formula:C₁₂H₁₄O₄
MDL Number:MFCD00009190