1,10-Phenanthroline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:66-71-7
:
Formula:C₁₂H₈N₂
MDL Number:MFCD00011678
:180.21 g/mol
Boiling Pt:>330 °C
Melting Pt:114…120 °C
Storage Temperature:Ambient
EINECS:200-629-2
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Frequently Bought Together