
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
Catalog # APOSOR2317-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:149.19 g/mol
Melting Pt:118…121 °C
:GHS07
Storage Temperature:Ambient
CAS Number:136030-00-7
Formula:C₉H₁₁NO
MDL Number:MFCD00216656