2-Furaldehyde
: 2-Furfurol, Furfural
:98-01-1
:
:C₅H₄O₂
:MFCD00003229
:159…162 °C
:–38 °C
:1.159 g/cm³ (25 °C)
:96.09 g/mol
:60 °C
:Ambient
:1199
:6.1+3 II
:13,04325
:202-627-7
:GHS02
:GHS06
:GHS08
- Environmentally Preferable:
Frequently Bought Together