
2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103788-65-4
:
Formula:C₁₂H₁₃NO₂
MDL Number:MFCD00100006
:203.24 g/mol
Melting Pt:72…75 °C
EINECS:452-560-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together