
4-Bromo-2,6-dichloroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:697-88-1
:
Formula:C₆H₄BrCl₂N
MDL Number:MFCD00025162
:240.91 g/mol
Melting Pt:85…86 °C
EINECS:211-809-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together