Order Entry
Ireland
ContactUsLinkComponent
 
Ethylene 99,5%
Catalog # APOSOR1710-80G
Supplier:  Apollo Scientific
CAS Number:  
Ethylene 99,5%
Catalog # APOSOR1710-80G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    80 g
  • Seq:
    3392
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Ethene
 :
 :28.05 g/mol
Flash Pt:–100 °C
EINECS:200-815-3
UN:1962
 :GHS02
 :GHS04
 :GHS07
Storage Temperature:Ambient
CAS Number:74-85-1
Formula:C₂H₄
MDL Number:MFCD00008604
Boiling Pt:–104 °C
Melting Pt:–169 °C
Density:0.97 g/cm³ (25 °C)
ADR:2.1
Merck Index:12,03837