
1-Boc-7-benzyloxy-3-hydroxymethylindole 98%
Catalog # APOSOR1707-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:353.42 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:914349-16-9
Formula:C₂₁H₂₃NO₄
MDL Number:MFCD05864726