


(±)-2-Aminobutan-1-ol
Catalog # APOSOR16670-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:89.14 g/mol
Boiling Pt:176…178 °C
Melting Pt:–2 °C
Density:0,943 g/cm³ (25 °C)
Flash Pt:84 °C
Storage Temperature:Ambient
EINECS:202-488-2
UN:2735
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:96-20-8
Formula:C₄H₁₁NO
MDL Number:MFCD00008095