(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, tert-Butyl ((1S,2R)-2-aminocyclohexyl)carbamate, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
CAS Number:365996-30-1
:
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD09952105
Molecular Weight:214.31 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together