
Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate
Catalog # APOSOR15504-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.23 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:105213-31-8
Formula:C₁₂H₁₂O₄
MDL Number:MFCD09864843