Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-phenyl-1,3-thiazol-4-ol
2-Amino-5-phenyl-1,3-thiazol-4-ol
2-Amino-5-phenyl-1,3-thiazol-4-ol
APOSOR15127-100MG
:
Apollo Scientific
:
2-Amino-5-phenyl-1,3-thiazol-4-ol
APOSOR15127-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
192.24 g/mol
:
240…242 °C
:
GHS07
:
Refrigerator
:
98879-58-4
:
C₉H₈N₂OS
:
MFCD08444003