Order Entry
Ireland
ContactUsLinkComponent
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Irritant
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Catalog # APOSOR14408-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Catalog # APOSOR14408-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.27 g/mol
Melting Pt:256…258 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:13906-08-6
Formula:C₁₀H₁₀N₂OS
MDL Number:MFCD00126093