Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-5-bromoindole-2-boronic acid
1-Boc-5-bromoindole-2-boronic acid
1-Boc-5-bromoindole-2-boronic acid
1-Boc-5-bromoindole-2-boronic acid
APOSOR14252-250MG
:
Apollo Scientific
:
1-Boc-5-bromoindole-2-boronic acid
APOSOR14252-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
339.98 g/mol
:
GHS07
:
Refrigerator
:
475102-13-7
:
C₁₃H₁₅BBrNO₄
:
MFCD03701683