5-Chloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:924869-18-1
:
Formula:C₁₄H₉ClN₂OS
MDL Number:MFCD08689693
:288.76 g/mol
Melting Pt:292…294 °C
Storage Temperature:Ambient
Frequently Bought Together