Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Diphenylethylamine
2,2-Diphenylethylamine 96%
2,2-Diphenylethylamine 96%
2,2-Diphenylethylamine 96%
APOSOR12742-25G
:
Apollo Scientific
:
2,2-Diphenylethylamine 96%
APOSOR12742-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
197.28 g/mol
:
48…49 °C
:
113 °C
:
Ambient
:
3963-62-0
:
C₁₄H₁₅N
:
MFCD00008143