
5-Chlorothiophene-2-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7283-96-7
:
Formula:C₅H₃ClOS
MDL Number:MFCD00047090
:146.59 g/mol
Boiling Pt:99 °C/21mmHg
Density:1,376 g/cm³ (25 °C)
Flash Pt:98 °C
EINECS:230-708-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together