Methyl-4-(4-chlorophenyl)-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:952183-64-1
:
Formula:C₁₅H₁₅ClN₂O₅
MDL Number:MFCD09817438
Melting Pt:164…165 °C
:338.75 g/mol
Storage Temperature:Ambient
Frequently Bought Together