Order Entry
Ireland
ContactUsLinkComponent
3-(4-Nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Irritant
3-(4-Nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Catalog # APOSOR12155-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Nitrophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Catalog # APOSOR12155-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.31 g/mol
Melting Pt:319…320 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:72176-80-8
Formula:C₁₄H₉N₃O₃S
MDL Number:MFCD02916354