
N-Methylpyrrole-2-boronic acid pinacol ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:850567-47-4
:
Formula:C₁₁H₁₈BNO₂
MDL Number:MFCD06659917
:207.08 g/mol
Melting Pt:44…50 °C
Flash Pt:82 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together