Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate
Catalog # APOSOR110974-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.25 g/mol
Storage Temperature:Ambient
CAS Number:916791-97-4
Formula:C₁₀H₁₆N₂O₂
MDL Number:MFCD00173782