
meso-Tetraphenylporphine
Catalog # APOSOR11095-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:614.75 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:917-23-7
Formula:C₄₄H₃₀N₄
MDL Number:MFCD00011680