Order Entry
Ireland
ContactUsLinkComponent
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester
Irritant
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester
Catalog # APOSOR110691-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester
Catalog # APOSOR110691-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.24 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:13996-05-9
Formula:C₈H₁₀N₂O₃S
MDL Number:MFCD06411556