
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:761446-44-0
:
Formula:C₁₀H₁₇BN₂O₂
MDL Number:MFCD03789259
:208.07 g/mol
Melting Pt:59…64 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together