
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17455-13-9
:
Formula:C₁₂H₂₄O₆
MDL Number:MFCD00005113
:264.32 g/mol
Boiling Pt:116 °C/0,2mmHg
Melting Pt:37…45 °C
Density:1,175 g/cm³ (25 °C)
Flash Pt:109 °C
EINECS:241-473-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together