
2-Chloro-N-methoxy-N-methylacetamide
Catalog # APOSOR10843-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:37…42 °C
Flash Pt:104 °C
:137.56 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:67442-07-3
Formula:C₄H₈ClNO₂
MDL Number:MFCD00134232