1,3-Dimethyl-2-imidazolidinone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80-73-9
:
Formula:C₅H₁₀N₂O
MDL Number:MFCD00003188
:114.15 g/mol
Boiling Pt:224…226 °C
Melting Pt:8 °C
Density:1,056 g/cm³ (25 °C)
Flash Pt:95 °C
EINECS:201-304-8
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together