
L(+)-Diethyl tartrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:87-91-2
:
Formula:C₈H₁₄O₆
MDL Number:MFCD00009143
:206.2 g/mol
Boiling Pt:280 °C
Melting Pt:17 °C
Density:1,204 g/cm³ (25 °C)
Flash Pt:93 °C
EINECS:201-783-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together