5-Cyano-1H-imidazole-4-carboxamide
Catalog # APOSOR0309-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:136.11 g/mol
UN:3439
:GHS06
Storage Temperature:Ambient
CAS Number:5372-23-6
Formula:C₅H₄N₄O
MDL Number:MFCD00833412
Melting Pt:276…278 °C
ADR:6.1 III