Order Entry
Ireland
ContactUsLinkComponent
1-Nonene
1-Nonene
Catalog # APOSOR01832-5ML
Supplier:  Apollo Scientific
CAS Number:  
1-Nonene
Catalog # APOSOR01832-5ML
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
Boiling Pt:146…147 °C
Melting Pt:–81 °C
Density:0,73 g/cm³ (25 °C)
Flash Pt:24 °C
Storage Temperature:Ambient
EINECS:204-681-7
UN:3295
ADR:3,III
 :126.24 g/mol
 :GHS02
 :GHS07
 :GHS08
CAS Number:124-11-8
Formula:C₉H₁₈
MDL Number:MFCD00009568