L(+)-Potassium sodium tartrate tetrahydrate
Catalog # APOSIN2941-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:282.22 g/mol
Melting Pt:70…80 °C
Density:1,79 g/cm³ (25 °C)
EINECS:206-156-8
:GHS07
Storage Temperature:Ambient
CAS Number:6381-59-5
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989