

Cesium nitrate 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7789-18-6
:
Formula:CsNO₃
MDL Number:MFCD00010963
:194.91 g/mol
Melting Pt:414 °C
Density:3,685 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-146-8
UN:1451
ADR:5.1,III
:GHS03
:GHS07
Frequently Bought Together