
Calcium bromide, anhydrous 99,5%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7789-41-5
:
Formula:Br₂Ca
MDL Number:MFCD00010902
:199.89 g/mol
Boiling Pt:806…812 °C
Melting Pt:730 °C (dec.)
Density:3,353 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-164-6
:GHS07
Frequently Bought Together