Order Entry
Ireland
ContactUsLinkComponent
BIX2188 Z-Isomer
BIX2188 Z-Isomer
Catalog # APOSBISN0195-1MG
Supplier:  Apollo Scientific
CAS Number:  
BIX2188 Z-Isomer
Catalog # APOSBISN0195-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 mg
  • Seq:
    2070
  • Environmentally Preferable:

Specifications

About this item

Synonyms: BIX-02188, BIX02188, (Z)-3-(((3-((dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide
 :
 :412.493 g/mol
Storage Temperature:Freezer
CAS Number:334949-59-6
 :1094614-84-2
Formula:C₂₅H₂₄N₄O₂
MDL Number:MFCD18157583