Order Entry
Ireland
ContactUsLinkComponent
Filgotinib (GLPG0634)
Filgotinib (GLPG0634)
Catalog # APOSBISN0129-25MG
Supplier:  Apollo Scientific
CAS Number:  
Filgotinib (GLPG0634)
Catalog # APOSBISN0129-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :425.5 g/mol
Storage Temperature:Ambient
CAS Number:1206161-97-8
Formula:C₂₁H₂₃N₅O₃S
MDL Number:MFCD20527867