

2-(2-Propynyloxy)ethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:122116-12-5
:
Formula:C₅H₉NO
MDL Number:MFCD16713446
:99.13 g/mol
Density:0,96 g/cm³ (25 °C)
UN:2734
ADR:8+3,II
:GHS02
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together