Neomycin sulphate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1405-10-3
:MFCD00077833
Formula:C₂₃H₄₆N₆O₁₃·3H₂SO₄
MDL Number:MFCD00150827
:908.87 g/mol
EINECS:215-773-1
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together