1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:41085-99-8
:
Formula:C₅₉H₉₇ClN₂O₄
MDL Number:MFCD00142354
:933.885 g/mol
Melting Pt:68 °C
Storage Temperature:Refrigerator
Frequently Bought Together