
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP red)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6769-80-8
:
Formula:C₈H₆BrClNO₄P·C₇H₉N
MDL Number:MFCD00210023
:GHS07
Storage Temperature:Freezer
Frequently Bought Together