Order Entry
Ireland
ContactUsLinkComponent
Rhesus macaque recombinant properdin (from HEK293 cells)
Rhesus macaque recombinant properdin (from HEK293 cells)
Catalog # ACRBPRN-R52H6-1MG
Supplier:  ACRO Biosystems
CAS Number:  
Rhesus macaque recombinant properdin (from HEK293 cells)
Catalog # ACRBPRN-R52H6-1MG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rhesus monkey
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    F7CGG5-1
  • Protein synonyms:
    CFP|PFC|Complement factor P|Properdin
  • Protein/peptide name:
    Properdin
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    50,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Rhesus macaque Properdin protein, His Tag.