Mouse recombinant CD27 ligand (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human CD27 at 1 μg/ml (100 µl/well) can bind Mouse CD27 Ligand, Mouse IgG2a Fc Tag, low endotoxin with a linear range of 0,3-2 ng/ml (QC tested).
- Gene ID:O55237-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD27LG|TNFSF7|TNFSF7G|CD27L|CD70
- Protein/peptide name:CD27 Ligand
- Purity:>95%
- Molecular weight:43,5 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together