Human recombinant ROR2 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:A1L4F5-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:NTRKR2|ROR2
- Protein/peptide name:ROR2
- Purity:>95%
- Molecular weight:43,2 kDa
- Endotoxin level:Low
- Formulation:Lyopilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together