Order Entry
Ireland
ContactUsLinkComponent
Human recombinant prolactin R (from HEK293)
Human recombinant prolactin R (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant prolactin R (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human Prolactin, Mouse IgG2a Fc Tag, low endotoxin at 5 μg/ml (100 µl/well) can bind Human Prolactin R, His Tag with a linear range of 20-156 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human GH, Tag Free at 5 μg/ml (100 µl/well) can bind Human Prolactin R, His Tag with a linear range of 0,313-5 μg/ml (Routinely tested).
  • Gene ID:
    P16471-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    PRL-R|Prolactin R|PRLR
  • Protein/peptide name:
    Prolactin R
  • Purity:
    >95%
  • Molecular weight:
    26,3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyopilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together