Order Entry
Ireland
ContactUsLinkComponent
Human recombinant PD-L1 (from HEK293)
Human recombinant PD-L1 (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human recombinant PD-L1 (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,01 EU per μg by the LAL method.
  • Biological activity:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Human PD-1, His Tag at 1 μg/ml (100 µl/well) can bind Human PD-L1, Llama IgG2b Fc Tag, low endotoxin with a linear range of 0,078-2,5 μg/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human PD-L1, Llama IgG2b Fc Tag, low endotoxin at 2 μg/ml (100 µl/well) can bind Biotinylated Human PD-1, Fc Tag with a linear range of 20-313 ng/ml (Routinely tested).
  • Gene ID:
    Q9NZQ7-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • Protein/peptide name:
    PD-L1
  • Purity:
    >95%
  • Molecular weight:
    53,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyopilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together