Human recombinant IL-8 (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P10145-1
- Protein synonyms:MDNCF|LYNAP|IL8|NAP-1|GCP1|MONAP|LECT|NAF|CXCL8|LUCT
- Protein/peptide name:IL-8
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:34,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together