Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus recombinant M-CSF R (from HEK293)
Cynomolgus recombinant M-CSF R (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Cynomolgus recombinant M-CSF R (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus M-CSF R, His Tag at 5 μg/ml (100 μl/well) can bind Biotinylated Human M-CSF, His,Avitag with a linear range of 2-16 ng/ml (QC tested)
  • Gene ID:
    G7P8P1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD115|CSFR|M-CSFR|C-FMS|FMS|FIM2|CSF1R
  • Protein/peptide name:
    M-CSF R
  • Purity:
    >95%
  • Molecular weight:
    57,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together