Cynomolgus recombinant M-CSF R (from HEK293)
Catalog # ACRBCSR-H5258-1MG
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Pack type:1 mg
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human M-CSF, His,Avitag at 2 μg/ml (100 μl/well) on streptavidin precoated (0,2 μg/well) plate, can bind Cynomolgus M-CSF R, Fc Tag with a linear range of 1-39 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human M-CSF, His Tag at 5 μg/ml (100 μl/well) can bind Cynomolgus M-CSF R, Fc Tag with a linear range of 1-39 ng/ml (Routinely tested)
- Gene ID:G7P8P1
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD115|CSFR|M-CSFR|C-FMS|FMS|FIM2|CSF1R
- Protein/peptide name:M-CSF R
- Purity:>95%
- Molecular weight:81,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation
- Nuclease-free:Y
- Shipping temperature:Powder,RT