Human recombinant complement component C2 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Complement Component C2 Protein, His Tag (MALS verified) (active enzyme), ACROBiosystems.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag (MALS verified) (active enzyme)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Gene ID:P06681-1
- Protein synonyms:Complement Component C2|Complement C2|C2
- Protein/peptide name:Complement Component C2
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:83,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together