Order Entry
Ireland
ContactUsLinkComponent
 
Human recombinant Fc gamma RI/ CD64 (from HEK293 cells)
Catalog # ACRBFCA-H52H1100UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human recombinant Fc gamma RI/ CD64 (from HEK293 cells)
Catalog # ACRBFCA-H52H1100UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Seq:
    02
  • Tag sequence:
    His Tag (MALS verified)
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Human CD64, His Tag (Cat. No. FCA-H52H1) captured on CM5 chip via anti-His antibody can bind Herceptin® with an affinity constant of 4.92 nM as determined in a SPR assay (Biacore 8K) (QC tested). Loaded Human CD64, His Tag (Cat. No. FCA-H52H1) on HIS1K Biosensor, can bind Herceptin with an affinity constant of 6.98 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Herceptin on Protein A Biosensor, can bind Human CD64, His Tag (Cat. No. FCA-H52H1) with an affinity constant of 0.297 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Herceptin on FAB2G Biosensor, can bind Human CD64, His Tag (Cat. No. FCA-H52H1) with an affinity constant of 0.244 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P12314-1
  • Protein synonyms:
    FCGR1|FCG1|FCRI|FCGR1A|IGFR1|CD64|CD64A
  • Protein/peptide name:
    Fc gamma RI/ CD64
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    31,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Human Fc gamma RI / CD64 Protein, His Tag (MALS verified), ACROBiosystems.