Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus recombinant glypican 3 (from HEK293)
Cynomolgus recombinant glypican 3 (from HEK293)
Catalog # ACRBGP3-C5225100UG
Supplier:  ACRO Biosystems
CAS Number:  
Cynomolgus recombinant glypican 3 (from HEK293)
Catalog # ACRBGP3-C5225100UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pack type:
    100 µg
  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Cynomolgus Glypican 3, His Tag at 0,5 μg/ml (100 μl/well) can bind Monoclonal Anti-Human GPC3 Antibody, Human IgG1 with a linear range of 0,2-3 ng/ml (QC tested)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    GPC3|SGBS|SGB|Glypican-3|SDYS |MXR7|SGBS1|OCI5|GTR2-2|DGSX
  • Protein/peptide name:
    Glypican 3
  • Purity:
    >85%
  • Molecular weight:
    38,1 kDa and 24,6 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item